Assay Details
| Gene Symbol: REXO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278120.1 | NP_001265049.1 | 3 - 4 | 328 | 81 | |
| GenBank mRNA | BM491337.1 | - | 3 - 4 | 328 | 81 | |
| BX929496.2 | - | 3 - 4 | 262 | 81 | ||
More Information
Set Membership: |
|

REXO2
Genomic Map