Assay Details
Gene Symbol: APOPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015287849.2 | XP_015143335.1 | 2 - 3 | 326 | 68 | |
XM_025151092.1 | XP_025006860.1 | 2 - 3 | 417 | 68 | ||
XM_025151093.1 | XP_025006861.1 | 2 - 3 | 415 | 68 | ||
XM_025151094.1 | XP_025006862.1 | 2 - 3 | 416 | 68 | ||
GenBank mRNA | BX929835.2 | - | 2 - 3 | 300 | 68 | |
CR353689.1 | - | 1 - 2 | 415 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
