Assay Details
| Gene Symbol: BST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001200043.1 | NP_001186972.1 | 4 - 5 | 585 | 75 | |
| XM_015285754.2 | XP_015141240.1 | 4 - 5 | 715 | 75 | ||
| XM_015285755.2 | XP_015141241.1 | 4 - 5 | 715 | 75 | ||
| GenBank mRNA | BX930007.2 | - | 4 - 5 | 721 | 75 | |
| HM769339.1 | - | 4 - 5 | 585 | 75 | ||
More Information
Set Membership: |
|

BST1
Genomic Map