Assay Details
Gene Symbol: BEND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284547.2 | XP_015140033.1 | 2 - 3 | 303 | 85 | |
GenBank mRNA | BX932932.2 | - | 2 - 3 | 182 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015284547.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
