Assay Details
| Gene Symbol: CCDC92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945529.3 | XP_004945586.1 | 3 - 4 | 312 | 93 | |
| XM_004945530.3 | XP_004945587.1 | 3 - 4 | 314 | 93 | ||
| XM_025155654.1 | XP_025011422.1 | 22 - 23 | 16888 | 93 | ||
| XM_025155655.1 | XP_025011423.1 | 23 - 24 | 16985 | 93 | ||
| XM_415108.5 | XP_415108.2 | 23 - 24 | 17061 | 93 | ||
| GenBank mRNA | BX933081.2 | - | 4 - 5 | 448 | 93 | |
More Information
Set Membership: |
|

CCDC92
Genomic Map