Assay Details
| Gene Symbol: NSUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938314.3 | XP_004938371.1 | 2 - 3 | 299 | 63 | |
| XM_015297960.2 | XP_015153446.1 | 2 - 3 | 300 | 63 | ||
| XM_416643.6 | XP_416643.3 | 2 - 3 | 303 | 63 | ||
| GenBank mRNA | BX934649.2 | - | 1 - 2 | 119 | 63 | |
More Information
Set Membership: |
|

NSUN3
Genomic Map