Assay Details
| Gene Symbol: RAB11FIP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286057.2 | XP_015141543.1 | 4 - 5 | 4145 | 66 | |
| XM_015286058.2 | XP_015141544.1 | 4 - 5 | 3878 | 66 | ||
| XM_015286059.2 | XP_015141545.1 | 4 - 5 | 3799 | 66 | ||
| XM_420890.6 | XP_420890.5 | 4 - 5 | 4146 | 66 | ||
| GenBank mRNA | BX934766.2 | - | 2 - 3 | 395 | 66 | |
More Information
Set Membership: |
|

RAB11FIP5
Genomic Map