Assay Details
Gene Symbol: NRBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199533.1 | NP_001186462.1 | 1 - 2 | 97 | 91 | |
GenBank mRNA | BX950839.2 | - | 1 - 2 | 97 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
