Assay Details
Gene Symbol: NRBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199533.1 | NP_001186462.1 | 3 - 4 | 221 | 106 | |
GenBank mRNA | BX950839.2 | - | 3 - 4 | 221 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
