Assay Details
| Gene Symbol: CPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416596.6 | XP_416596.1 | 4 - 5 | 830 | 69 | |
| GenBank mRNA | CR354187.1 | - | 4 - 5 | 518 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPOX| Gene Symbol: CPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416596.6 | XP_416596.1 | 4 - 5 | 830 | 69 | |
| GenBank mRNA | CR354187.1 | - | 4 - 5 | 518 | 69 | |
Set Membership: |
|