Assay Details
| Gene Symbol: TM2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424392.6 | XP_424392.1 | 1 - 2 | 229 | 79 | |
| GenBank mRNA | CR386237.1 | - | 1 - 2 | 238 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM2D2| Gene Symbol: TM2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424392.6 | XP_424392.1 | 1 - 2 | 229 | 79 | |
| GenBank mRNA | CR386237.1 | - | 1 - 2 | 238 | 79 | |
Set Membership: |
|