Assay Details
| Gene Symbol: DTX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286836.2 | XP_015142322.1 | 6 - 7 | 1585 | 78 | |
| GenBank mRNA | AF184236.1 | - | 5 - 6 | 727 | 78 | |
More Information
Set Membership: |
|

DTX4
Genomic Map