Assay Details
| Gene Symbol: NR3C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159345.1 | NP_001152817.1 | 5 - 6 | 2911 | 70 | |
| XM_015276099.2 | XP_015131585.1 | 6 - 7 | 2641 | 70 | ||
| XM_015276101.2 | XP_015131587.1 | 6 - 7 | 2840 | 70 | ||
| XM_015276102.2 | XP_015131588.1 | 6 - 7 | 2555 | 70 | ||
| XM_025149459.1 | XP_025005227.1 | 6 - 7 | 2641 | 70 | ||
| XM_025149460.1 | XP_025005228.1 | 5 - 6 | 2160 | 70 | ||
| GenBank mRNA | AF468211.1 | - | 1 - 2 | 76 | 70 | |
| FJ808618.1 | - | 5 - 6 | 2911 | 70 | ||
| GU230774.1 | - | 1 - 2 | 72 | 70 | ||
More Information
Set Membership: |
|

NR3C2
Genomic Map