Assay Details
| Gene Symbol: ZBTB38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937067.3 | XP_004937124.1 | 2 | 9588 | 91 | |
| XM_015276973.2 | XP_015132459.1 | 2 | 3663 | 91 | ||
| XM_015276974.2 | XP_015132460.1 | 2 | 3521 | 91 | ||
| XM_015276975.2 | XP_015132461.1 | 3 | 3988 | 91 | ||
| XM_015276976.2 | XP_015132462.1 | 2 | 3851 | 91 | ||
| XM_025153694.1 | XP_025009462.1 | 4 | 3806 | 91 | ||
| XM_025153695.1 | XP_025009463.1 | 4 | 4251 | 91 | ||
| XM_025153696.1 | XP_025009464.1 | 3 | 3774 | 91 | ||
| XM_025153697.1 | XP_025009465.1 | 3 | 3617 | 91 | ||
| XM_422601.6 | XP_422601.3 | 3 | 3853 | 91 | ||
| GenBank mRNA | BX929416.1 | - | 1 | 547 | 91 | |
| BX932404.1 | - | 1 | 694 | 91 | ||
More Information
Set Membership: |
|

ZBTB38
Genomic Map