Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277440.1 | NP_001264369.1 | 2 - 3 | 228 | 112 | |
| XM_015276926.2 | XP_015132412.1 | 2 - 3 | 303 | 112 | ||
| XM_025153356.1 | XP_025009124.1 | 2 - 3 | 162 | 112 | ||
| GenBank mRNA | BX929424.1 | - | 2 - 3 | 190 | 112 | |
| CN237805.1 | - | 2 - 3 | 228 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277440.1, XM_015276926.2, XM_025153356.1 transcripts.
Set Membership: |
|

CEP19
Genomic Map