Assay Details
Gene Symbol: TMEM267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_424781.6 | XP_424781.1 | 2 - 3 | 278 | 84 | |
GenBank mRNA | BX932048.1 | - | 2 - 3 | 198 | 84 |
Gene Symbol: CZH5ORF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145052.1 | XP_025000820.1 | 13 - 14 | 5490 | 84 | |
XM_025145054.1 | XP_025000822.1 | 13 - 14 | 5490 | 84 | ||
XM_025145055.1 | XP_025000823.1 | 13 - 14 | 5490 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_424781.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
