Assay Details
| Gene Symbol: TMEM267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145057.1 | XP_025000825.1 | 2 - 3 | 405 | 103 | |
| XM_424781.6 | XP_424781.1 | 3 - 4 | 656 | 103 | ||
| GenBank mRNA | BX932048.1 | - | 3 - 4 | 576 | 103 | |
| Gene Symbol: CZH5ORF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145052.1 | XP_025000820.1 | 14 - 15 | 5868 | 103 | |
| XM_025145053.1 | XP_025000821.1 | 15 - 16 | 5977 | 103 | ||
More Information
Set Membership: |
|

TMEM267
Genomic Map