Assay Details
| Gene Symbol: GPX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277569.2 | XP_015133055.1 | 1 - 2 | 260 | 147 | |
| GenBank mRNA | BX932298.1 | - | 1 - 2 | 195 | 147 | |
More Information
Set Membership: |
|

GPX8
Genomic Map