Assay Details
| Gene Symbol: ELP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277490.1 | NP_001264419.1 | 2 - 3 | 156 | 57 | |
| GenBank mRNA | BX932848.1 | - | 2 - 3 | 127 | 57 | |
| CD219096.1 | - | 2 - 3 | 159 | 57 | ||
| CR405721.1 | - | 2 - 3 | 133 | 57 | ||
| DR424316.1 | - | 2 - 3 | 104 | 57 | ||
More Information
Set Membership: |
|

ELP6
Genomic Map