Assay Details
| Gene Symbol: SLC9A3R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294453.2 | XP_015149939.1 | 5 - 6 | 710 | 73 | |
| XM_414851.6 | XP_414851.3 | 5 - 6 | 972 | 73 | ||
| GenBank mRNA | BX933578.1 | - | 626 | 73 | ||
More Information
Set Membership: |
|

SLC9A3R2
Genomic Map