Assay Details
| Gene Symbol: PIGW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946737.3 | XP_004946794.1 | 2 | 638 | 88 | |
| XM_015296151.2 | XP_015151637.1 | 2 | 651 | 88 | ||
| GenBank mRNA | BX935190.1 | - | 1 | 851 | 88 | |
More Information
Set Membership: |
|

PIGW
Genomic Map