Assay Details
| Gene Symbol: NECAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277413.1 | NP_001264342.1 | 7 - 8 | 590 | 63 | |
| XM_025147097.1 | XP_025002865.1 | 8 - 9 | 826 | 63 | ||
| XM_025147098.1 | XP_025002866.1 | 7 - 8 | 769 | 63 | ||
| XM_025147099.1 | XP_025002867.1 | 8 - 9 | 703 | 63 | ||
| XM_025147100.1 | XP_025002868.1 | 7 - 8 | 781 | 63 | ||
| XM_025147101.1 | XP_025002869.1 | 6 - 7 | 642 | 63 | ||
| XM_025147102.1 | XP_025002870.1 | 6 - 7 | 727 | 63 | ||
| XM_025147103.1 | XP_025002871.1 | 6 - 7 | 560 | 63 | ||
| GenBank mRNA | BX936064.1 | - | 7 - 8 | 589 | 63 | |
| CN219471.1 | - | 7 - 8 | 590 | 63 | ||
More Information
Set Membership: |
|

NECAB1
Genomic Map