Assay Details
Gene Symbol: CNTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293181.2 | XP_015148667.1 | 9 - 10 | 1195 | 65 | |
XM_025154515.1 | XP_025010283.1 | 9 - 10 | 1196 | 65 | ||
XM_414435.6 | XP_414435.2 | 9 - 10 | 1196 | 65 | ||
GenBank mRNA | X85809.1 | - | 179 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
