Assay Details
Gene Symbol: DDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282047.4 | NP_001268976.1 | 9 - 10 | 1374 | 68 | |
XM_015298004.2 | XP_015153490.1 | 9 - 10 | 1411 | 68 | ||
XM_025143188.1 | XP_024998956.1 | 9 - 10 | 1338 | 68 | ||
GenBank mRNA | AJ720165.1 | - | 9 - 10 | 1376 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
