Assay Details
| Gene Symbol: PGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006303.1 | NP_001006303.1 | 5 - 6 | 538 | 77 | |
| XM_025142522.1 | XP_024998290.1 | 4 - 5 | 616 | 77 | ||
| GenBank mRNA | AJ720644.1 | - | 5 - 6 | 538 | 77 | |
More Information
Set Membership: |
|

PGD
Genomic Map