Assay Details
Gene Symbol: SLITRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640550.4 | XP_003640598.1 | 2 | 1092 | 106 | |
GenBank mRNA | CR389928.1 | - | 1 | 747 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
