Assay Details
| Gene Symbol: RPL35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277457.1 | NP_001264386.1 | 4 - 5 | 388 | 94 | |
| XM_025153370.1 | XP_025009138.1 | 4 - 5 | 398 | 94 | ||
| GenBank mRNA | BU255532.1 | - | 4 - 5 | 354 | 94 | |
| BU328575.1 | - | 4 - 5 | 388 | 94 | ||
| BX929952.1 | - | 1 - 2 | 513 | 94 | ||
| CR390693.1 | - | 3 - 4 | 615 | 94 | ||
More Information
Set Membership: |
|

RPL35A
Genomic Map