Assay Details
| Gene Symbol: TCERG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421827.6 | XP_421827.4 | 13 | 2803 | 92 | |
| GenBank mRNA | CR391756.1 | - | 1 | 546 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCERG1L| Gene Symbol: TCERG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421827.6 | XP_421827.4 | 13 | 2803 | 92 | |
| GenBank mRNA | CR391756.1 | - | 1 | 546 | 92 | |
Set Membership: |
|