Assay Details
| Gene Symbol: LRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282979.2 | XP_015138465.1 | 3 - 4 | 753 | 77 | |
| XM_418378.6 | XP_418378.4 | 4 - 5 | 809 | 77 | ||
| GenBank mRNA | CR406808.1 | - | 1 - 2 | 36 | 77 | |
More Information
Set Membership: |
|

LRP12
Genomic Map