Assay Details
Gene Symbol: C9ORF58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415461.6 | XP_415461.1 | 1 - 2 | 101 | 77 | |
GenBank mRNA | CR523365.1 | - | 1 - 2 | 60 | 77 | |
KJ101611.1 | - | 1 - 2 | 66 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
