Assay Details
| Gene Symbol: PCDHGC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY325274.1 | - | 2 | 2848 | 56 | |
| Gene Symbol: LOC100857975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154946.1 | XP_025010714.1 | 1 | 4081 | 56 | |
| Gene Symbol: LOC100858040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154949.1 | XP_025010717.1 | 2 | 4194 | 56 | |
| Gene Symbol: LOC100858937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293558.2 | XP_015149044.1 | 1 | 2825 | 56 | |
| Gene Symbol: LOC100859022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154947.1 | XP_025010715.1 | 1 | 2412 | 56 | |
| Gene Symbol: LOC107054480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293559.2 | XP_015149045.1 | 1 | 6325 | 56 | |
| Gene Symbol: LOC107054481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293560.2 | XP_015149046.1 | 1 | 2289 | 56 | |
| Gene Symbol: LOC107054485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293563.2 | XP_015149049.2 | 1 | 2676 | 56 | |
| Gene Symbol: LOC107054486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293564.2 | XP_015149050.1 | 1 | 2825 | 56 | |
| Gene Symbol: LOC107054487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154944.1 | XP_025010712.1 | 1 | 2612 | 56 | |
| Gene Symbol: LOC107054488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293566.2 | XP_015149052.1 | 1 | 2796 | 56 | |
| Gene Symbol: LOC107056921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015301880.2 | XP_015157366.1 | 1 | 7324 | 56 | |
| Gene Symbol: LOC112533402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154941.1 | XP_025010709.1 | 1 | 2664 | 56 | |
| Gene Symbol: LOC112533403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154943.1 | XP_025010711.1 | 1 | 2627 | 56 | |
| Gene Symbol: LOC427618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154948.1 | XP_025010716.1 | 1 | 2503 | 56 | |
| Gene Symbol: PCDHGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245037.1 | NP_001231966.1 | 2 | 2848 | 56 | |
| XM_004950675.3 | XP_004950732.3 | 1 | 3239 | 56 | ||
| Gene Symbol: PCDHGA2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154942.1 | XP_025010710.1 | 1 | 2801 | 56 | |
More Information
Set Membership: |
|

PCDHGC3
Genomic Map