Assay Details
Gene Symbol: YARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006314.1 | NP_001006314.1 | 10 - 11 | 1101 | 64 | |
XM_025142997.1 | XP_024998765.1 | 10 - 11 | 1095 | 64 | ||
GenBank mRNA | AJ720626.1 | - | 10 - 11 | 1101 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
