Assay Details
| Gene Symbol: LSM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006337.2 | NP_001006337.1 | 2 - 3 | 167 | 72 | |
| XM_025144262.1 | XP_025000030.1 | 2 - 3 | 278 | 72 | ||
| XM_025144263.1 | XP_025000031.1 | 2 - 3 | 151 | 72 | ||
| XM_025144265.1 | XP_025000033.1 | 2 - 3 | 278 | 72 | ||
| GenBank mRNA | AJ719882.1 | - | 2 - 3 | 168 | 72 | |
| CR385825.1 | - | 1 - 2 | 99 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025144262.1, XM_025144263.1, XM_025144265.1 transcripts.
Set Membership: |
|

LSM7
Genomic Map