Assay Details
| Gene Symbol: LSM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006337.2 | NP_001006337.1 | 3 - 4 | 235 | 91 | |
| XM_025144262.1 | XP_025000030.1 | 3 - 4 | 346 | 91 | ||
| XM_025144263.1 | XP_025000031.1 | 3 - 4 | 219 | 91 | ||
| XM_025144264.1 | XP_025000032.1 | 2 - 3 | 1597 | 91 | ||
| XM_025144265.1 | XP_025000033.1 | 3 - 4 | 346 | 91 | ||
| GenBank mRNA | AJ719882.1 | - | 3 - 4 | 236 | 91 | |
| CR385825.1 | - | 2 - 3 | 167 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025144262.1, XM_025144263.1, XM_025144264.1, XM_025144265.1 transcripts.
Set Membership: |
|

LSM7
Genomic Map