Assay Details
| Gene Symbol: FAM206A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006383.1 | NP_001006383.1 | 5 - 6 | 410 | 96 | |
| XM_015282375.2 | XP_015137861.1 | 5 - 6 | 581 | 96 | ||
| XM_025147280.1 | XP_025003048.1 | 6 - 7 | 593 | 96 | ||
| GenBank mRNA | AJ721017.1 | - | 5 - 6 | 410 | 96 | |
More Information
Set Membership: |
|

FAM206A
Genomic Map