Assay Details
Gene Symbol: ABCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006440.1 | NP_001006440.1 | 4 - 5 | 460 | 68 | |
GenBank mRNA | AJ720308.1 | - | 4 - 5 | 460 | 68 | |
AM049247.1 | - | 3 - 4 | 345 | 68 | ||
AM050269.1 | - | 3 - 4 | 345 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
