Assay Details
Gene Symbol: NSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204565.1 | NP_989896.1 | 3 - 4 | 296 | 137 | |
GenBank mRNA | AF498103.1 | - | 3 - 4 | 296 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
