Assay Details
Gene Symbol: MEPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204569.1 | NP_989900.1 | 1 - 2 | 87 | 96 | |
XM_015276110.2 | XP_015131596.1 | 1 - 2 | 793 | 96 | ||
GenBank mRNA | AF148716.3 | - | 1 - 2 | 87 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
