Assay Details
Gene Symbol: IL2RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204596.1 | NP_989927.1 | 4 - 5 | 740 | 72 | |
GenBank mRNA | AF143806.1 | - | 4 - 5 | 740 | 72 | |
AY772091.1 | - | 4 - 5 | 740 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
