Assay Details
Gene Symbol: NKX2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204616.1 | NP_989947.1 | 1 - 2 | 388 | 75 | |
XM_015287273.2 | XP_015142759.1 | 2 - 3 | 3185 | 75 | ||
XM_015287274.2 | XP_015142760.1 | 2 - 3 | 4448 | 75 | ||
XM_025150545.1 | XP_025006313.1 | 3 - 4 | 4527 | 75 | ||
XM_025150546.1 | XP_025006314.1 | 3 - 4 | 3264 | 75 | ||
XM_025150547.1 | XP_025006315.1 | 2 - 3 | 426 | 75 | ||
GenBank mRNA | AF110995.1 | - | 1 - 2 | 298 | 75 | |
AJ223618.1 | - | 1 - 2 | 388 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
