Assay Details
| Gene Symbol: HOXA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204619.1 | NP_989950.1 | 1 - 2 | 695 | 141 | |
| GenBank mRNA | JF317540.1 | - | 1 - 2 | 650 | 141 | |
| M81250.1 | - | 1 - 2 | 695 | 141 | ||
More Information
Set Membership: |
|

HOXA11
Genomic Map