Assay Details
| Gene Symbol: OPN4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204625.3 | NP_989956.3 | 1 - 2 | 142 | 109 | |
| XM_015276115.2 | XP_015131601.1 | 11 - 12 | 2968 | 109 | ||
| XM_015276116.2 | XP_015131602.1 | 11 - 12 | 7932 | 109 | ||
| XM_015276120.2 | XP_015131606.1 | 3 - 4 | 1021 | 109 | ||
| XM_025149493.1 | XP_025005261.1 | 11 - 12 | 8006 | 109 | ||
| XM_025149494.1 | XP_025005262.1 | 9 - 10 | 8036 | 109 | ||
| XM_025149495.1 | XP_025005263.1 | 11 - 12 | 5144 | 109 | ||
| XM_025149496.1 | XP_025005264.1 | 10 - 11 | 8010 | 109 | ||
| XM_025149497.1 | XP_025005265.1 | 11 - 12 | 7997 | 109 | ||
| XM_025149498.1 | XP_025005266.1 | 10 - 11 | 7822 | 109 | ||
| GenBank mRNA | AY036061.1 | - | 1 - 2 | 144 | 109 | |
| EU124630.1 | - | 1 - 2 | 144 | 109 | ||
| EU124631.1 | - | 1 - 2 | 144 | 109 | ||
More Information
Set Membership: |
|

OPN4-1
Genomic Map