Assay Details
| Gene Symbol: OPN4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204625.3 | NP_989956.3 | 8 - 9 | 1241 | 131 | |
| XM_015276115.2 | XP_015131601.1 | 18 - 19 | 4067 | 131 | ||
| XM_015276116.2 | XP_015131602.1 | 18 - 19 | 9031 | 131 | ||
| XM_015276120.2 | XP_015131606.1 | 10 - 11 | 2120 | 131 | ||
| XM_025149493.1 | XP_025005261.1 | 18 - 19 | 9105 | 131 | ||
| XM_025149494.1 | XP_025005262.1 | 16 - 17 | 9135 | 131 | ||
| XM_025149495.1 | XP_025005263.1 | 18 - 19 | 6243 | 131 | ||
| XM_025149496.1 | XP_025005264.1 | 17 - 18 | 9109 | 131 | ||
| XM_025149497.1 | XP_025005265.1 | 18 - 19 | 9096 | 131 | ||
| XM_025149498.1 | XP_025005266.1 | 17 - 18 | 8921 | 131 | ||
| GenBank mRNA | AY036061.1 | - | 8 - 9 | 1243 | 131 | |
| EU124630.1 | - | 8 - 9 | 1243 | 131 | ||
More Information
Set Membership: |
|

OPN4-1
Genomic Map