Assay Details
| Gene Symbol: AKR1B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204629.2 | NP_989960.1 | 1 - 2 | 118 | 94 | |
| GenBank mRNA | AJ295030.1 | - | 1 - 2 | 94 | ||
| BU297526.1 | - | 1 - 2 | 102 | 94 | ||
| BU385394.1 | - | 1 - 2 | 103 | 94 | ||
More Information
Set Membership: |
|

AKR1B10
Genomic Map