Assay Details
| Gene Symbol: ADCY9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204630.2 | NP_989961.2 | 8 - 9 | 2820 | 62 | |
| XM_015294128.2 | XP_015149614.1 | 7 - 8 | 2741 | 62 | ||
| GenBank mRNA | AJ401469.1 | - | 8 - 9 | 2793 | 62 | |
More Information
Set Membership: |
|

ADCY9
Genomic Map