Assay Details
| Gene Symbol: MINPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204644.1 | NP_989975.1 | 1 - 2 | 616 | 121 | |
| GenBank mRNA | U59421.1 | - | 1 - 2 | 616 | 121 | |
More Information
Set Membership: |
|

MINPP1
Genomic Map