Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 12 - 13 | 1612 | 102 | |
| XM_003641367.4 | XP_003641415.1 | 14 - 15 | 2376 | 102 | ||
| XM_015286526.2 | XP_015142012.1 | 14 - 15 | 2602 | 102 | ||
| XM_015286527.2 | XP_015142013.1 | 15 - 16 | 2556 | 102 | ||
| XM_015286528.2 | XP_015142014.1 | 10 - 11 | 1942 | 102 | ||
| GenBank mRNA | AB046524.1 | - | 12 - 13 | 1612 | 102 | |
More Information
Set Membership: |
|

OVOA
Genomic Map