Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 17 - 18 | 2306 | 131 | |
| XM_003641367.4 | XP_003641415.1 | 19 - 20 | 3070 | 131 | ||
| XM_015286526.2 | XP_015142012.1 | 19 - 20 | 3296 | 131 | ||
| XM_015286527.2 | XP_015142013.1 | 20 - 21 | 3250 | 131 | ||
| XM_015286528.2 | XP_015142014.1 | 15 - 16 | 2636 | 131 | ||
| GenBank mRNA | AB046524.1 | - | 17 - 18 | 2306 | 131 | |
More Information
Set Membership: |
|

OVOA
Genomic Map