Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 19 - 20 | 2560 | 102 | |
| XM_003641367.4 | XP_003641415.1 | 21 - 22 | 3324 | 102 | ||
| XM_015286526.2 | XP_015142012.1 | 21 - 22 | 3550 | 102 | ||
| XM_015286527.2 | XP_015142013.1 | 22 - 23 | 3504 | 102 | ||
| XM_015286528.2 | XP_015142014.1 | 17 - 18 | 2890 | 102 | ||
| GenBank mRNA | AB046524.1 | - | 19 - 20 | 2560 | 102 | |
More Information
Set Membership: |
|

OVOA
Genomic Map