Assay Details
| Gene Symbol: OVOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204661.1 | NP_989992.1 | 23 - 24 | 3224 | 92 | |
| XM_003641367.4 | XP_003641415.1 | 25 - 26 | 3988 | 92 | ||
| XM_015286526.2 | XP_015142012.1 | 25 - 26 | 4214 | 92 | ||
| XM_015286527.2 | XP_015142013.1 | 26 - 27 | 4168 | 92 | ||
| XM_015286528.2 | XP_015142014.1 | 21 - 22 | 3554 | 92 | ||
| GenBank mRNA | AB046524.1 | - | 23 - 24 | 3224 | 92 | |
More Information
Set Membership: |
|

OVOA
Genomic Map